Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2702400

Sigma-Aldrich

Anti-N-cadherin antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

99kDa

species reactivity

zebrafish

concentration

1mg/mL

technique(s)

immunohistochemistry: 1:100-1:500
western blot: 1:5,000-1:20,000

isotype

IgG

shipped in

wet ice

Storage temp.

−20°C

General description

N-cadherin, also known as cadherin 2 (CDH2), is encoded by the gene mapped to human chromosome 18q11.2. N-Cadherin belongs to the cadherin family. It is highly expressed on the cell membrane of endothelium, nervous and muscular cells.

Immunogen

Recombinant protein encompassing a sequence within the center region of zebrafish N-cadherin.

Application

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-Wm: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Cadherins plays a vital role in cell-cell adhesion. N-glycosylation of N-cadherin is associated with the process of tumorigenesis especially in mammary gland, renal system and ovary. In addition, N-glycosylation helps in maintaining N-cadherin stability and facilitates glioma migration. Over expression of CDH2 gene was observed in tumor cells. It is known to induce the invasive and chemoresistant attributes to the cancer cells. Loss of expression of the gene was observed in adrenocortical carcinoma. Thus, it can be used as a molecular biomarker in various types of tumoral adrenocortical tissue.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Purified by antigen-affinity chromatography

Physical form

1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endothelial cell-to-cell junctions
Dejana E, et al.
Faseb Journal, 9(10), 910-918 (1995)
Structure of the human N-cadherin gene: YAC analysis and fine chromosomal mapping to 18q11.2.
Wallis J, et al.
Genomics, 22(1), 172-179 (1994)
Chemotherapy enriches for an invasive triple-negative breast tumor cell subpopulation expressing a precursor form of N-cadherin on the cell surface.
Nelson ER, et al.
Oncotarget, 7(51), 84030-84030 (2016)
Telomerase and N-Cadherin Differential Importance in Adrenocortical Cancers and Adenomas.
Pereira SS, et al.
Journal of Cellular Biochemistry, 118(8), 2064-2071 (2017)
N-Glycosylation at Asn 402 Stabilizes N-Cadherin and Promotes Cell-Cell Adhesion of Glioma Cells.
Xu Y, et al.
Journal of Cellular Biochemistry, 118(6), 1423-1431 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service