Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2702371

Sigma-Aldrich

Monoclonal Anti-Cep 164 antibody produced in mouse

clone 17, affinity isolated antibody

Synonym(s):

NPHP15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

17, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: 1:500-1:20000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEP164(22897)

Immunogen

The immunogen used to generate this antibody corresponds to Cep 164

Application

Suggested starting dilutions are as follows: WB: 1:500-1:20000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

This monoclonal antibody binds to Cep164 or centrosonal protein 164kDa. Cep164 plays a role in microtubule organization and/or maintenance for the formation of primary cilia (PC), a microtubule-based structure that protrudes from the surface of epithelial cells. Plays a critical role in G2/M checkpoint and nuclear divisions. A key player in the DNA damage-activated ATR/ATM signaling cascade since it is required for the proper phosphorylation of H2AX, RPA,CHK2 and CHK1. Plays a critical role in chromosome segregation, acting as a mediator required for the maintenance of genomic stability through modulation of MDC1, RPA and CHK1. [from UniProt]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Affinity purified by Protein G

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service