SAB2702198
Monoclonal Anti-GFP antibody produced in mouse
clone GT859, affinity isolated antibody
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
biological source
mouse
conjugate
peroxidase conjugate
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
GT859, monoclonal
form
buffered aqueous solution
mol wt
29kDa
concentration
0.36mg/mL
technique(s)
western blot: 1:5000-1:20000
isotype
IgG2b
shipped in
wet ice
storage temp.
−20°C
Immunogen
Full length GFP recombinant protein
Application
Suggested starting dilutions are as follows: WB: 1:5000-1:20000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Other Notes
Purification: Affinity purified by Protein G
Physical form
1%BSA, 0.05M Tris, 0.15M NaCl (pH7.4). 0.005% Thimerosal was added as a preservative.
Disclaimer
For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.


Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Inga S Shchelik et al.
Acta biomaterialia, 136, 99-110 (2021-10-04)
Biohybrid microswimmers exploit the natural abilities of motile microorganisms e.g. in releasing cargo on-demand. However, using such engineered swarms to release antibiotics addressing bacterial infections has not yet been realized. Herein, a design strategy for biohybrid microswimmers is reported, which
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service