Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2702164

Sigma-Aldrich

Monoclonal Anti-NCOA1 antibody produced in mouse

Synonym(s):

RIP160, SRC1, bHLHe42, bHLHe74

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1135
monoclonal

form

liquid

species reactivity

monkey, human, rat, mouse

concentration

1 mg/mL

technique(s)

ChIP: suitable
immunoprecipitation (IP): 0.5-2 μg/mL
western blot: 1:500-1:3000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

human ... NCOA1(8648)

Immunogen

Amino acids 477-947 of human SRC-1.

Application

For IP: Use at a concentration of 0.5 - 2 μg/mL. For WB: 1:500-1:3000. Predicted molecular weight: 169 kDa. Optimal dilutions/concentrations should be determined by the researcher.

Biochem/physiol Actions

Clone 1135 recognizes SRC-1, steroid-receptor coactivator-1. SRC-1 is a member of a gene family that functions in the full transcriptional activation of the steroid hormone receptor superfamily and contains intrinsic histone acetyltransferase activity. SRC-1 has a molecular weight of 160 kDa and has a basic helix-loop-helix (bHLH)-PAS domain.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service