Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2702151

Sigma-Aldrich

Monoclonal Anti-ID3 antibody produced in mouse

Synonym(s):

Hlh462, bHLHb25, inhibitor of DNA binding 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$512.00

$512.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$512.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$512.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B11
monoclonal

form

liquid

mol wt

14 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ID3(15903)

Immunogen

The full-length (amino acids 1-119) mouse Id3 gene expressed in E. coli.

application

For ICC/IF: 1:100-1:1000. For IP: Use at a concentration of 1 - 5 μg/mL. For WB: Use at a concentration of 1 - 5 μg/mL. Predicted molecular weight: 13 kDa. Optimal dilutions/cocentrations should be determined by the researcher.

Biochem/physiol Actions

Id3-2B11 recognizes Id3, a 14a protein that is a member of the Id family of helix-loop-helix proteins. Id proteins are believed to be dominant negative regulators of bHLH proteins involved in cell differentiation and implicated in the control of cell growth, cellular differentiation, apoptosis and carcinogenesis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, pH 7.4, containing no preservatives

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service