Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2702058

Sigma-Aldrich

Anti-LAMP2 antibody produced in rabbit

Synonym(s):

CD107b, LAMP-2, LAMPB, LGP110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

45 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 500-3000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LAMP2(3920)

Immunogen

Recombinant fragment corresponding to a region within amino acids 73 and 339 of LAMP2 (Uniprot ID#P13473)

Application

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

The protein encoded by this gene is a member of a family of membrane glycoproteins. This glycoprotein provides selectins with carbohydrate ligands. It may play a role in tumor cell metastasis. It may also function in the protection, maintenance, and adhesion of the lysosome. Alternative splicing of this gene results in multiple transcript variants encoding distinct proteins. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su-shan Luo et al.
Clinical neuropathology, 33(4), 284-291 (2014-04-03)
Danon disease is an Xlinked dominant lysosomal glycogen storage disorder characterized by cardiomyopathy, skeletal myopathy, and mental retardation. This study described two Chinese cases of Danon disease in order to broaden the phenotypic and genetic spectrum. Clinical data were collected
Asako Kurauchi-Mito et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 37(9), 830-835 (2014-05-17)
The (pro)renin receptor ((P)RR) is known to play an important role in the pathogenesis of vascular complications in diabetes mellitus and hypertension through its function in activating the local renin-angiotensin system. Recent studies have shown that the (P)RR is an
Makiko Kihara et al.
PloS one, 9(7), e103928-e103928 (2014-08-01)
Itm2a is a type II transmembrane protein with a BRICHOS domain. We investigated the temporospatial mRNA and protein expression patterns of Itm2a in the developing lower first molar, and examined the subcellular localization of Itm2a in murine dental epithelial (mDE6)
Sandro Alves et al.
Acta neuropathologica, 128(5), 705-722 (2014-05-27)
There is still no treatment for polyglutamine disorders, but clearance of mutant proteins might represent a potential therapeutic strategy. Autophagy, the major pathway for organelle and protein turnover, has been implicated in these diseases. To determine whether the autophagy/lysosome system
Sudhakar Raja Subramaniam et al.
Neurobiology of disease, 70, 204-213 (2014-07-13)
Parkinson's disease (PD) is characterized by the progressive degeneration of nigrostriatal dopaminergic neurons leading to motor deficits. The mechanisms underlying the preferential vulnerability of nigrostriatal dopaminergic neurons in PD remain poorly understood. Recent evidence supports a role for mitochondrial dysfunction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service