Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2701992

Sigma-Aldrich

Anti-EPAS1 antibody produced in rabbit

Synonym(s):

HLF, MOP2, PASD2, bHLHe73

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$512.00

$512.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$512.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$512.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

96 kDa

species reactivity

human, zebrafish

concentration

0.37 mg/mL

technique(s)

ChIP: suitable
western blot: 1:500-1:3000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EPAS1(2034)

Immunogen

Recombinant fragment corresponding to a region within amino acids 74 and 376 of HIF2 alpha (Uniprot ID#Q99814)

Application

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ChIP assay: Assay-dependent dilution. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

This gene encodes a transcription factor involved in the induction of genes regulated by oxygen, which is induced as oxygen levels fall. The encoded protein contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. Mutations in this gene are associated with erythrocytosis familial type 4. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service