Skip to Content
MilliporeSigma
All Photos(11)

Key Documents

SAB2701973

Sigma-Aldrich

Anti-SOX2 antibody produced in rabbit

Synonym(s):

ANOP3, MCOPS3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

34 kDa

species reactivity

mouse, human, zebrafish

concentration

1 mg/mL

technique(s)

flow cytometry: 50-200
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: 500-3000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SOX2(6657)

Immunogen

Recombinant fragment corresponding to a region within amino acids 1 and 158 of SOX2 (Uniprot ID#P48431)

Application

Suggested starting dilutions are as follows: FACS: 1:50-1:200, ICC/IF: 1:100-1:1000, IHC-Fr: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

This intronless gene encodes a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of this gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. Mutations in this gene have been associated with optic nerve hypoplasia and with syndromic microphthalmia, a severe form of structural eye malformation. This gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai-xia Lu et al.
Neurological research, 36(3), 207-214 (2014-02-12)
Efforts have been made by tissue engineers to create a permissive environment for neural regeneration, and to enhance the efficiency of neural stem cell (NSC) transplantation. However, to acquire sufficient number of seed cells on the material appears to be
Xiangxin Lou et al.
Neuroscience letters, 579, 1-6 (2014-07-13)
The presence of stem cells in the organ of Corti raises the hope of regeneration of mammalian inner ear cells. However, little is known about the distribution of endogenous stem cells in the inner ear as well as their sphere-forming
Irem Dogan et al.
Lung cancer (Amsterdam, Netherlands), 85(1), 1-6 (2014-04-22)
Primary and acquired resistance to EGFR TKIs in EGFR mutant lung cancer occurs primarily through secondary mutations in EGFR or Met amplification. Drug resistance can also be mediated by expression of pluripotency transcription factors, such as OCT4, SOX2 and NANOG
Ana Benito-Gonzalez et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(38), 12865-12876 (2014-09-19)
Mechano-sensory hair cells (HCs), housed in the inner ear cochlea, are critical for the perception of sound. In the mammalian cochlea, differentiation of HCs occurs in a striking basal-to-apical and medial-to-lateral gradient, which is thought to ensure correct patterning and
Changhwan Yoon et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(15), 3974-3988 (2014-06-21)
Gastric cancers may harbor a subset of cells with cancer stem cell (CSC) properties, including chemotherapy resistance, and CD44 is a gastric CSC marker. The Hedgehog (HH) pathway is a key developmental pathway that can be subverted by CSCs during

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service