Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2701832

Sigma-Aldrich

Anti-hsp90ab1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

hsp90ab1, hsp90b, hsp90beta, wu:fa29f01

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

zebrafish

technique(s)

immunohistochemistry: suitable
western blot: 1:5000-1:20000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

zebrafish ... hsp90ab1(30573)

Immunogen

Recombinant protein containing a sequence corresponding to a region within amino acids 47 and 213 of hsp90ab1 according to NP_571385

Application

Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-Wm: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Molecular chaperone. Has ATPase activity.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui-Yu Chen et al.
Veterinary parasitology, 205(3-4), 540-550 (2014-10-02)
Anisakid nematodes are distributed worldwide in a wide variety of marine fishes and they are known to cause the zoonotic disease, anisakiasis. The temperature control is commonly applied for prevention and control of anisakiasis. To analyze the cellular response to
Balázs Rada et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 63(10), 821-830 (2014-07-23)
We studied the involvement of calcium and calcium-activated NADPH oxidases in NLRP3 inflammasome activation and IL-1β release to better understand inflammasome signaling in macrophages. Human volunteer blood donors were recruited to isolate monocytes to differentiate them into macrophages. Wild-type or
Xin Zhao et al.
Naunyn-Schmiedeberg's archives of pharmacology, 387(11), 1079-1089 (2014-08-12)
Arsenic trioxide (As2O3) is used to treat acute promyelocytic leukemia. However, the cardiotoxicity of long QT syndrome restricts its clinical application. Previous studies showed that As2O3 can damage the human ether-a-go-go-related gene (hERG) current via disturbing its trafficking to cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service