SAB2701753
Anti-pygb antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
Synonym(s):
phosphorylase, glycogen; brain, pygb
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
zebrafish
technique(s)
immunohistochemistry: suitable
western blot: suitable
NCBI accession no.
shipped in
wet ice
Storage temp.
−20°C
Gene Information
zebrafish ... pygb(403051)
Immunogen
Features and Benefits
Disclaimer

Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service