Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2701743

Sigma-Aldrich

Anti-lcp1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

L-plastin, cb245, lcp1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

zebrafish

technique(s)

immunohistochemistry: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

zebrafish ... lcp1(30583)

Immunogen

Recombinant protein containing a sequence corresponding to a region within amino acids 104 and 354 of lcp1 according to NP_571395

Application

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000, IHC-Wm: 1:50-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Actin-binding protein. Plays a role in the activation of T-cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jolien Van Houcke et al.
NPJ Regenerative medicine, 8(1), 31-31 (2023-06-17)
The young African turquoise killifish has a high regenerative capacity, but loses it with advancing age, adopting several aspects of the limited form of mammalian regeneration. We deployed a proteomic strategy to identify pathways that underpin the loss of regenerative
Rom Keshet et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16365-16370 (2014-11-05)
Adipocyte differentiation, or adipogenesis, is a complex and highly regulated process. A recent proteomic analysis has predicted that the nonreceptor tyrosine kinase Abelson murine leukemia viral oncogene (c-Abl) is a putative key regulator of adipogenesis, but the underlying mechanism remained
Helen L Barrett et al.
PloS one, 9(8), e104826-e104826 (2014-08-15)
Infants of women with gestational diabetes mellitus (GDM) are more likely to be born large for gestational age with a higher percentage body fat. Elevated maternal lipids may contribute to this. Placental lipases such as lipoprotein lipase (LPL), endothelial lipase
Shu-Han Miao et al.
CNS neuroscience & therapeutics, 20(8), 754-762 (2014-03-19)
To investigate the role of astrocytic JWA expression in dopaminergic (DA) neuron degeneration and in the pathogenesis of Parkinson's disease (PD). Conditional astrocytic JWA null (JWA∆2/∆2/GFAP-Cre) mice and U251 glioma cells were used to evaluate the effects of JWA gene
Yanbo Yang et al.
Biochemical and biophysical research communications, 451(4), 632-636 (2014-08-19)
Lipoprotein lipase (LPL) is an extracellular lipase that primarily hydrolyzes triglycerides within circulating lipoproteins. Macrophage LPL contributes to atherogenesis, but the mechanisms behind it are poorly understood. We hypothesized that the products of lipoprotein hydrolysis generated by LPL promote atherogenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service