Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2701319

Sigma-Aldrich

Anti-PNP antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

FLJ94043, FLJ97288, FLJ97312, NP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: 1:100-1:1,000
immunohistochemistry: suitable
western blot: 1:500-1:3,000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

human ... PNP(4860)

Immunogen

Recombinant fragment corresponding to a region within amino acids 83 and 289 of Nucleoside phosphorylase according to AAI04207

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC: Assay-dependent dilution, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

This gene encodes an enzyme which reversibly catalyzes the phosphorolysis of purine nucleosides. The enzyme is trimeric, containing three identical subunits. Mutations which result in nucleoside phosphorylase deficiency result in defective T-cell (cell-mediated) immunity but can also affect B-cell immunity and antibody responses. Neurologic disorders may also be apparent in patients with immune defects. A known polymorphism at aa position 51 that does not affect enzyme activity has been described. A pseudogene has been identified on chromosome 2. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anwar M Hashem et al.
Journal of immunology (Baltimore, Md. : 1950), 193(2), 722-734 (2014-06-15)
CD40L, a key regulator of the immune system, was studied as both a targeting ligand and a molecular adjuvant in nucleoprotein (NP)-based host defense against influenza in mouse models with different genetic backgrounds. Adenoviral vectors secreting NP-CD40L fusion protein (denoted
Aihua Song et al.
Antiviral research, 111, 60-68 (2014-09-15)
Influenza virus is a global health concern due to its unpredictable pandemic potential. Frequent mutations of surface molecules, hemagglutinin (HA) and neuraminidase (NA), contribute to low efficacy of the annual flu vaccine and therapeutic resistance to standard antiviral agents. The
Marie O Pohl et al.
Journal of virology, 88(19), 11271-11283 (2014-07-18)
Influenza A virus (IAV) entry is a multistep process that requires the interaction of the virus with numerous host factors. In this study, we demonstrate that prolidase (PEPD) is a cellular factor required by IAV for successful entry into target
Derek C Rookhuizen et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(31), E3224-E3233 (2014-07-24)
Recent studies have demonstrated important roles of nucleic acid-sensing Toll-like receptors (TLRs) in promoting protective antibody responses against several viruses. To dissect how recognition of nucleic acids by TLRs enhances germinal center (GC) responses, mice selectively deleted for myeloid differentiation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service