Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2701008

Sigma-Aldrich

Anti-CCL5 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

CCL5, D17S136E, MGC17164, RANTES

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1:500-1:3000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL5(6352)

Immunogen

Synthetic peptide corresponding to a region within amino acids 27 and 91 of RANTES according to NP_002976

Application

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

This gene is one of several CC cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongfeng Sun et al.
Transplantation, 97(12), 1233-1239 (2014-05-16)
Regulated on activation normal T-cell expressed and secreted (RANTES)-chemokine ligand 5 plays a key role in mediating heart transplant rejection. Suppression of RANTES-mediated signals can reduce leukocyte recruitment and mitigate transplant rejection severity. The present study describes the construction of
Hye Young Kim et al.
Cytokine, 68(1), 40-49 (2014-03-25)
An anti-inflammatory cytokine, interleukin-10 (IL-10) exerts inhibitory effects on vascular inflammation. Chemokines promote vascular inflammation and play a pathogenic role in the development and maintenance of hypertension. However, chemokine CCL5 has down-regulatory effects on angiotensin II (Ang II)-induced hypertensive mediators.
Egle Cekanaviciute et al.
Journal of immunology (Baltimore, Md. : 1950), 193(1), 139-149 (2014-05-27)
The balance between controlling infection and limiting inflammation is particularly precarious in the brain because of its unique vulnerability to the toxic effects of inflammation. Astrocytes have been implicated as key regulators of neuroinflammation in CNS infections, including infection with
Esteban Grasso et al.
The Journal of endocrinology, 221(1), 121-131 (2014-02-05)
During early pregnancy, the human uterus undergoes profound tissue remodeling characterized by leukocyte invasion and production of proinflammatory cytokines, followed by tissue repair and tolerance maintenance induction. Vasoactive intestinal peptide (VIP) is produced by trophoblast cells and modulates the maternal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service