Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB2700205

Sigma-Aldrich

Anti-APLNR (N-terminal) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

AGTRL1, APJ, APJR, APLNR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: 500-3000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... APLNR(187)

Immunogen

Synthetic peptide corresponding to a region within amino acids 2 and 96 of Apelin Receptor according to NP_005152

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

There are at least two distinct receptor subtypes of angiotensin II: angiotensin II receptor, type 1 (AGTR1) and angiotensin II receptor, type 2 (AGTR2). Most of the effects of angiotensin II are mediated by the AGTR1 receptor. This gene is related to the AGTR1 gene by sequence similarity. It was cloned based on a conserved transmembrane domain found in members of the G protein-coupled receptor (GPCR) gene family. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danai Georgiadou et al.
Scientific reports, 9(1), 19077-19077 (2019-12-15)
Preeclampsia is a frequent gestational hypertensive disorder with equivocal pathophysiology. Knockout of peptide hormone ELABELA (ELA) has been shown to cause preeclampsia-like symptoms in mice. However, the role of ELA in human placentation and whether ELA is involved in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service