Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SAB2700198

Sigma-Aldrich

Anti-CXCR7 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

CMKOR1, CXCR7, GPR159, RDC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 500-3000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CXCR7(57007)

Immunogen

Synthetic peptide corresponding to a region within amino acids 187 and 282 of CXCR7 according to NP_064707

Application

Suggested starting dilutions are as follows: FACS: Assay-dependent dilution, ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, IP: Assay-dependent dilution, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

This gene encodes a member of the G-protein coupled receptor family. Although this protein was earlier thought to be a receptor for vasoactive intestinal peptide (VIP), it is now considered to be an orphan receptor, in that its endogenous ligand has not been identified. The protein is also a coreceptor for human immunodeficiency viruses (HIV). Translocations involving this gene and HMGA2 on chromosome 12 have been observed in lipomas. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Meng et al.
Renal failure, 46(1), 2300727-2300727 (2024-01-08)
Renal fibrosis is a common feature of various chronic kidney diseases. However, the underlying mechanism remains poorly understood. The CXC chemokine receptor (CXCR) family plays a role in renal fibrosis; however, the detailed mechanisms have not been elucidated. In this
Hongyan Mo et al.
Antioxidants & redox signaling, 27(6), 345-362 (2016-12-15)
Oxidative stress plays a role in mediating podocyte injury and proteinuria. However, the underlying mechanism remains poorly understood. In this study, we investigated the potential role of C-X-C chemokine receptor type 4 (CXCR4), the receptor for stromal cell-derived factor 1α
Kina Miyoshi et al.
Human pathology, 45(9), 1900-1909 (2014-08-05)
CXC chemokine receptor 4 (CXCR4) expression is reportedly correlated with both vascular endothelial growth factor (VEGF) expression and poor prognosis in a variety of cancers. Its relation to CXC chemokine receptor 7 (CXCR7) is also noted in several malignancies, including
Tsung-Han Wu et al.
Cellular immunology, 290(1), 107-115 (2014-06-25)
We developed an in vitro model to evaluate the effect of products secreted from different colorectal cancer (CRC) cell lines on specific phenotypic switching and functional alterations in THP-1 cells. We co-cultured the human monocytic cell line, THP-1, or phorbol-12-myristate-13-acetate
Hans Jürgen Wester et al.
Theranostics, 5(6), 618-630 (2015-04-01)
Chemokine ligand-receptor interactions play a pivotal role in cell attraction and cellular trafficking, both in normal tissue homeostasis and in disease. In cancer, chemokine receptor-4 (CXCR4) expression is an adverse prognostic factor. Early clinical studies suggest that targeting CXCR4 with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service