Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB2700196

Sigma-Aldrich

Anti-Envelope protein antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Dengue virus 2 Envelop protein, Envelop protein (Dengue virus 2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

Dengue virus

technique(s)

immunofluorescence: suitable
western blot: 5000-20000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Immunogen

Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 495 of Envelope protein (Dengue virus 2) according to NP_739583

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:2000, WB: 1:5000-1:20000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruisheng Wang et al.
Analytical chemistry, 91(8), 5415-5423 (2019-03-22)
In this work, we demonstrate a rapid diagnostic platform with potential to transform clinical diagnosis of acute febrile illnesses in resource-limited settings. Acute febrile illnesses such as dengue and chikungunya, which pose high burdens of disease in tropical regions, share
Raúl G Saraiva et al.
PLoS neglected tropical diseases, 12(4), e0006443-e0006443 (2018-04-26)
Dengue virus (DENV) is the most prevalent and burdensome arbovirus transmitted by Aedes mosquitoes, against which there is only a limited licensed vaccine and no approved drug treatment. A Chromobacterium species, C. sp. Panama, isolated from the midgut of A.
Erika R Schwarz et al.
Viruses, 11(9) (2019-09-01)
Zika virus (ZIKV) is a vertically and sexually transmissible virus resulting in severe congenital malformation. The goal of this study was to develop an ovine model of ZIKV infection. Between 28-35 days gestation (DG), four pregnant animals were infected with
Chontida Tangsongcharoen et al.
Biochimica et biophysica acta. Proteins and proteomics, 1867(2), 89-97 (2018-11-06)
The recent emergence of Zika virus (ZIKV) has caused global concern as a result of the association with neurological disorders, and brain development dysfunction in fetuses of mothers who become infected with ZIKV during pregnancy. The NS2B-NS3 protease is important
Hua Lu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(6), 6801-6812 (2019-02-28)
Herpes simplex virus 1 (HSV-1) is a contagious neurotropic herpesvirus responsible for oral lesions and herpesviral encephalitis. The HSV-1 envelope contains N-glycosylated proteins involved in infection and that are candidate drug targets. NGI-1 is a small-molecule inhibitor of oligosaccharyltransferase (OST)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service