Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SAB2700024

Sigma-Aldrich

Anti-NS4B antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Dengue virus 2 Nonstructural protein NS4B, Non-structure protein 4B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

Dengue virus

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
western blot: 1000-10000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Immunogen

Synthetic peptide containing a sequence corresponding to a region within amino acids 1 and 14 of Non-structural protein 4B (Dengue virus 2 ) according to NP_739589

Application

Suggested starting dilutions are as follows: FACS: Assay-dependent dilution, ICC/IF: 1:100-1:2000, WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 40% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service