Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501988

Sigma-Aldrich

Anti-Smgc antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

2310010P21Rik, DXImx49e, Muc19, Sfc21, submandibular gland protein C

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$586.00

$586.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$586.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$586.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... Smgc(223809)

General description

Submandibular gland protein C (Smgc), also known as muc19 (mucin 19), belongs to the large gel-forming mucin family. It is secreted by the submandibular and sublingual glands in rodents. Smgc gene is mapped to mouse chromosome 15E3. The gene contains 60 exons and spans ~105kb on genomic DNA.

Immunogen

Peptide with sequence C-KLEPKYENPTNGS, from the internal region of the protein sequence according to NP_945121.1

Biochem/physiol Actions

Submandibular gland protein C (Smgc) might play an important role in the cyto-differentiation of mucous and seromucous acinar cells. It also acts as a marker for tubule secretory cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular cloning and characterization of the neonatal rat and mouse submandibular gland protein SMGC.
Zinzen K M, et al.
Gene, 334, 23-33 (2004)
NFIB regulates embryonic development of submandibular glands.
Mellas R E, et al.
Journal of Dental Research, 94(2), 312-319 (2015)
Tissue Distibution of Murine Muc19/Smgc Gene Products
Das B, et al.
The Journal of Histochemistry and Cytochemistry, 58(2), 141-156 (2010)
Samantha D Praktiknjo et al.
Nature communications, 11(1), 991-991 (2020-02-23)
Characterizing the complex composition of solid tumors is fundamental for understanding tumor initiation, progression and metastasis. While patient-derived samples provide valuable insight, they are heterogeneous on multiple molecular levels, and often originate from advanced tumor stages. Here, we use single-cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service