Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501848

Sigma-Aldrich

Anti-IDH3B antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

FLJ11043, H-IDHB, MGC903, isocitrate dehydrogenase 3 (NAD+) beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

canine, mouse, rat, pig, human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IDH3B(3420)

General description

Isocitrate dehydrogenase 3 (NAD+) β (IDH3B) is a subunit of IDH3. IDH3 catalyzes the oxidation of isocitrate to α-ketoglutarate. The gene encoding this protein is localized on human chromosome 20p13. Absence of isocitrate dehydrogenase 3 (NAD+) β (IDH3B) has been linked to retinitis pigmentosa.

Immunogen

Peptide with sequence C-HAASRSQAEDVRVE, from the internal region of the protein sequence according toNP_008830.2; NP_777280.1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genes and mutations causing retinitis pigmentosa
Daiger SP
Clinical Genetics (2013)
Insights from retinitis pigmentosa into the roles of isocitrate dehydrogenases in the Krebs cycle.
Hartong DT
Nature Genetics (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service