Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501718

Sigma-Aldrich

Anti-COPS3 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

COP9 complex subunit 3, COP9 constitutive photomorphogenic homolog subunit 3 (Arabidopsis), CSN3, SGN3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$586.00

$586.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$586.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$586.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, canine, mouse, rat, bovine

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COPS3(8533)

General description

COPS3 (COP9 signalosome subunit 3) encodes the third subunit of an eight-subunit complex, the COP9 signalosome (CSN), and is located on chromosome 17p11.2.

Immunogen

Peptide with sequence C-SLYKKNIQRLT, from the internal region of the protein sequence according toNP_003644.2

Application

Anti-COPS3 antibody produced in goat is suitable for indirect ELISA and western blot applications.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong-sheng Yu et al.
Cancer chemotherapy and pharmacology, 69(5), 1173-1180 (2012-01-13)
Csn3 (or CSN3) encodes the third subunit of an eight-subunit complex, the COP9 signalosome (CSN), which acts as a protein kinase and a deneddylase in mammalian cells. Previous studies have shown that Csn3 is essential for maintenance of cell proliferation
Joeri Both et al.
PloS one, 7(1), e30907-e30907 (2012-02-01)
Osteosarcoma is the most common primary malignancy of bone. The tumours are characterized by high genomic instability, including the occurrence of multiple regions of amplifications and deletions. Chromosome region 17p11.2-p12 is amplified in about 25% of cases. In previous studies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service