Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501574

Sigma-Aldrich

Anti-JAK2 (C-terminal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Janus kinase 2 (a protein tyrosine kinase), Tyrosine-protein kinase JAK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable
indirect ELISA: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JAK2(3717)

General description

Janus kinase 2 (JAK2) encodes a tyrosine kinase which belongs to the Jak family of protein kinases. The gene encoding this protein is localized on human chromosome 9p24.

Immunogen

Peptide with sequence RVDQVRDNMAG, from the C-terminal region of the protein sequence according to AAC23653.1

Biochem/physiol Actions

Janus kinase 2 (JAK2) plays an important role in the activities of cytokine receptors. Overexpression of JAK2 results in neuroblastoma. Mutations in the gene have been linked to primary resistance to PD-1 (anti-programmed death protein 1) blockade therapy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jak2 Is Essential for Signaling through a Variety of Cytokine Receptors
Evan Parganas
Cell, 93(3), 385- 395 (1998)
Hodgkin's lymphoma cell lines are characterized by frequent aberrations on chromosomes 2p and 9p including REL and JAK2?
Stefan Joos
International Journal of Cancer. Journal International Du Cancer, 103(4) , 489-495 (2003)
Cancer-Associated Fibroblasts Share Characteristics and Protumorigenic Activity with Mesenchymal Stromal Cells.
Borriello L
Cancer Research, 77(18), 5142-5157 (2017)
Primary Resistance to PD-1 Blockade Mediated by JAK1/2 Mutations.
Shin DS
Cancer Discovery, 7(2), 188-201 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service