Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501553

Sigma-Aldrich

Anti-Itga11 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-4732459H24Rik, Anti-Itga11, Integrin a11, Integrin alpha-11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Itga11(319480)

General description

Integrin subunit α 11 (ITGA11) is expressed in stromal fibroblasts. Together with the integrin subunit β, it forms a receptor for collagen.

Immunogen

Peptide with sequence PTEEDLSHAPQRNH, from the internal region (near C-terminus) of the protein sequence according to NP_795896.4

Biochem/physiol Actions

Integrin subunit α 11 (ITGA11) may have a role in the organization of interstitial collagen matrices. It has been shown to be upregulated in non-small-cell lung carcinoma. Mice, which are deficient in ITGA11, exhibit signs of dwarfism and malnutrition.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

alpha11beta1 integrin is a receptor for interstitial collagens involved in cell migration and collagen reorganization on mesenchymal nonmuscle cells.
Tiger CF
Developmental Biology (2001)
Integrin a11 regulates IGF2 expression in fibroblasts to enhance tumorigenicity of human non-small-cell lung cancer cells
Chang-Qi Zhu
Proceedings of the National Academy of Sciences of the USA (2007)
a11?1 Integrin-Dependent Regulation of Periodontal Ligament Function in the Erupting Mouse Incisor
Svetlana N
Molecular and Cellular Biology (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service