Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501318

Sigma-Aldrich

Anti-NEFM antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NEF3, Anti-NF-M, Anti-NFM, neurofilament, medium polypeptide

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEFM(4741)

General description

NEFM (neurofilament, medium polypeptide) is a highly available cytoskeleton intermediate filament (IF) protein in post-mitotic neurons. It is mapped to human chromosome 8q21. It is abundantly present in the nervous system. In human, the protein has 13 carboxy-terminal domains with phosphorylation sites.

Immunogen

Peptide with sequence CQASHITVERKDYLK, from the internal region of the protein sequence according to NP_005373.2; NP_001099011.1.

Application

Anti-NEFM antibody produced in goat is suitable for indirect ELISA and western blot analysis.

Biochem/physiol Actions

NEFM (neurofilament, medium polypeptide) protein is involved in the axonal transport and neuronal cell apoptosis. Phosphorylation of NEFM allows the association between NEFM and microtubules. Deregulated phosphorylation of NEFM is associated with Alzheimer disease (AD). From gene mutation analysis, it has been predicted that NEFM may be a susceptibility factor for the Parkinson′s disease (PD).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn L Penney et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 24(1), 255-260 (2014-11-06)
Numerous germline genetic variants are associated with prostate cancer risk, but their biologic role is not well understood. One possibility is that these variants influence gene expression in prostate tissue. We therefore examined the association of prostate cancer risk variants
Rejko Krüger et al.
Neuroscience letters, 351(2), 125-129 (2003-10-30)
Neurofilament M, a major component of Lewy bodies, represents an interesting candidate in the pathogenesis of Parkinson's disease (PD). We performed detailed mutation analyses of the NF-M gene in 322 familial and sporadic PD patients. Two polymorphisms (Ala475Thr and Gly697Arg)
Raul Perez-Olle et al.
Journal of neuropathology and experimental neurology, 63(7), 759-774 (2004-08-05)
The human neurofilament medium (hNFM) subunit is one of the 3 neurofilament (NF) polypeptides, which are the most abundant intermediate filament (IF) proteins in post-mitotic neurons. The formation of neurofilamentous aggregates is a pathological hallmark of many neurodegenerative diseases, including
Parvathi Rudrabhatla et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 24(11), 4396-4407 (2010-07-14)
Aberrant hyperphosphorylation of neuronal cytoskeletal proteins is one of the major pathological hallmarks of neurodegenerative disorders such as Alzheimer disease (AD), amyotrophic lateral sclerosis (ALS), and Parkinson's disease (PD). Human NF-M/H display a large number of multiple KSP repeats in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service