Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501250

Sigma-Aldrich

Anti-AGAP2 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CENTG1, Anti-FLJ16430, Anti-GGAP2, Anti-KIAA0167, ArfGAP with GTPase domain, ankyrin repeat and PH domain 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$586.00

$586.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$586.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$586.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AGAP2(116986)

Related Categories

General description

The gene AGAP2 (ArfGAP with GTPase domain, ankyrin repeat and PH domain 2) belongs to the group of proteins called AZAPs that contain a catalytic core of PH (pleckstrin homology), Arf GAP and two or more ankyrin repeat domains. The AZAP family is further divided into four subfamilies, of which AGAPs contain an NH2-terminal GTPase-like domain (GLD), a split PH domain, and the GAP domain followed by four ankyrin repeats. Agap2 is found to be overexpressed in various human cancers.

Immunogen

Peptide with sequence CQASLDSIREAVINSQ, from the internal region of the protein sequence according to NP_055585.1.

Biochem/physiol Actions

The gene AGAP2 (ArfGAP with GTPase domain, ankyrin repeat and PH domain 2) encodes an Arf GAP (GTPase activating protein) that catalyzes the hydrolysis of GTP bound to Arf (ADP-ribosylation factors). The AGAPs are involved in the regulation of membrane trafficking and remodeling of the actin cytoskeleton. AGAP2 is found to associate with the clathrin adaptor protein AP-1 (activator protein 1) and participate in endosomal trafficking that is dependent on AP-1/Rab4 (RAS-related GTP-binding protein). It is also involved in facilitating the invasion of glioblastoma cells. Agap2 interacts with FAK (focal adhesion kinase) to form a complex and increase its activity, thereby promoting focal adhesion disassembly, resulting in increased cell migration.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arf GTPase-activating protein AGAP2 regulates focal adhesion kinase activity and focal adhesion remodeling.
Zhu Y
The Journal of Biological Chemistry, 284, 13489-13496 (2009)
Arif A Surani et al.
iScience, 25(12), 105519-105519 (2022-11-26)
The relevance of minor transcription start sites in broad promoters is not well understood. We have studied AGAP2 expression in prostate cancer and chronic myeloid leukemia (CML), showing transcription is initiated from alternative transcription start sites (TSSs) within a single

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service