Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501197

Sigma-Aldrich

Anti-UBE2F (C-terminal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-MGC18120, Anti-NCE2, ubiquitin-conjugating enzyme E2F (putative)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$586.00

$586.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$586.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$586.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBE2F(140739)

General description

Ubiquitin conjugating enzyme E2 F (UBE2F), also known as NEDD8-conjugating enzyme 2 (NCE2) or MGC18120, is encoded by the gene mapped to human chromosome 2q37.3.

Immunogen

Peptide with sequence CHLRDKEDFRNKVDD, from the C-Terminus of the protein sequence according to NP_542409.1.

Biochem/physiol Actions

Ubiquitin conjugating enzyme E2 F (UBE2F) acts as a neural precursor cell expressed, developmentally down-regulated 8 (NEDD8)-conjugating enzyme E2 in the neddylation cascade. It activates the polyubiquitin synthesis of cullin RING (really interesting new gene) ligase viral infectivity factor (CRL5Vif)-core binding factor β (CBFβ) complex in vitro.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Role of the COP9 Signalosome and Neddylation in DNA Damage Signaling and Repair.
Chung d and Dellaire G
Biomolecules (2015)
What was the set of ubiquitin and ubiquitin-like conjugating enzymes in the eukaryote common ancestor?
Michelle C
Journal of Molecular Evolution (2009)
Inhibition of a NEDD8 Cascade Restores Restriction of HIV by APOBEC3G.
Stanley DJ
PLoS Pathogens (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service