Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

SAB2501128

Sigma-Aldrich

Anti-MS4A1 (C-terminal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-B1, Anti-Bp35, Anti-CD20, Anti-LEU-16, membrane-spanning 4-domains, subfamily A, member 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MS4A1(931)

General description

Membrane spanning 4-domains A1 (MS4A1), also referred to as cluster of differentiation 20 (CD20), is expressed on the surface of human B lymphocytes and on a small subset of T cells. The 33-35kDa, non-glycosylated protein has four membrane-spanning domains with both the amino and carboxy termini of the protein localized to the cytoplasm, and a short 43 residue-extracellular segment. It is part of the membrane-spanning 4A gene family. The gene encoding MS4A1 is localized on human chromosome 11q12-13.

Immunogen

Peptide with sequence CQDQESSPIENDSSP, from the C Terminus of the protein sequence according to NP_068769.2; NP_690605.1.

Biochem/physiol Actions

Membrane spanning 4-domains A1 (MS4A1) or CD20 participates in the development and cell cycle progression in B-cells. It has been found to interact with proteins like adapter protein p75/80, CD40 and major histocompatibility complex class II proteins (MHC II). It may participate in Ca2+ influx across plasma membranes, maintenance of Ca2+ concentration and activation of B-cells. CD20 activates Src leading to rapid phosphorylation of phospholipase C-γ1 (PLC-γ1) and PLC-γ2, in turn activating caspase-3 signaling of apoptosis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

From the bench to the bedside: ways to improve rituximab efficacy.
Carton G
Blood (2004)
Kristine Salmina et al.
Genes, 10(2) (2019-01-30)
Aneuploidy should compromise cellular proliferation but paradoxically favours tumour progression and poor prognosis. Here, we consider this paradox in terms of our most recent observations of chemo/radio-resistant cells undergoing reversible polyploidy. The latter perform the segregation of two parental groups

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service