Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501044

Sigma-Aldrich

Anti-TIRAP/MAL (Isoform B) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Adapter protein wyatt, Anti-Mal, Anti-Toll-interleukin 1 receptor (TIR) domain-containing adapter protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$586.00

$586.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$586.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$586.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TIRAP(114609)

General description

TIR domain containing adaptor protein (TIRAP) is an adaptor protein and the gene encoding it is localized on human chromosome 11q24.2.

Immunogen

Peptide with sequence C-EGEGERDSATVSDL from the C Terminus of the protein sequence according to NP_683708.1.

Biochem/physiol Actions

TIR domain containing adaptor protein (TIRAP) takes part in the toll-like receptor 4 (TLR4) signaling pathway. It activates various proteins and indirectly influences the release of inflammatory cytokines. This protein also has a role in the activation of proinflammatory genes.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of S180L polymorphism in etiology of malaria caused by Plasmodium falciparum in a small group of Pakistani population.
Nawaz SK
Bosnian Journal of Basic Medical Sciences / Udruzenje Basicnih Mediciniskih Znanosti = Association of Basic Medical Sciences (2015)
Genetic variants in the TIRAP gene are associated with increased risk of sepsis-associated acute lung injury.
Song Z
BMC Medical Genetics (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service