Peptide with sequence C-QAYDRQKNPSKEER from the internal region of the protein sequence according to NP_000449.1; NP_006472.1.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Aberrantly elevated expression in obesity of microRNAs (miRNAs), including the miRNA miR-802, contributes to obesity-associated metabolic complications, but the mechanisms underlying the elevated expression are unclear. Farnesoid X receptor (FXR), a key regulator of hepatic energy metabolism, has potential for
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.