Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2501016

Sigma-Aldrich

Anti-TAK1/MAP3K7 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Mitogen-activated protein kinase kinase kinase 7, Anti-TGF-β activated kinase 1, Anti-TGF1a, Anti-Transforming growth factor-β-activated kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP3K7(6885)

General description

MAP3K7 (mitogen-activated protein kinase kinase kinase 7) gene encodes a serine/threonine protein kinase that is ubiquitously expressed. It is activated by transforming growth factor-β (TGF-β) and BMP (bone morphogenetic protein) and mediates cellular functions such as transcription regulation and apoptosis. It functions in the p38 and janus kinase (JNK) signaling pathways and also plays an important role in balancing the BMP- and Wnt-signaling pathways. It mediates JNK pathway induced by environmental stresses. It is essential for nuclear factor κB (NF-κB) activation. MAP3K7 gene has been mapped to human chromosome 6q15. Deletions in this region have been observed in cases of prostate cancer indicating its role as a tumor suppressor.

Immunogen

Peptide with sequence C-AELDQDEKDQQNT from the internal region of the protein sequence according to NP_003179.1; NP_663304.1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genomic deletion of MAP3K7 at 6q12-22 is associated with early PSA recurrence in prostate cancer and absence of TMPRSS2:ERG fusions
Martina Kluth
Modern Pathology (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service