Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

SAB2500750

Sigma-Aldrich

Anti-PARK7/DJ-1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DJ1, Anti-Oncogene DJ1, Anti-Parkinson disease (autosomal recessive)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PARK7(11315)

Immunogen

Peptide with sequence C-AAQVKAPLVLKD from the C Terminus of the protein sequence according to NP_009193.

Application

Anti-PARK7/DJ-1 antibody produced in goat is suitable for indirect ELISA and western blot.

Biochem/physiol Actions

PARK7 is a member of novel glyoxalase family. It exerts mitochondrial activity. It has been suggested that PARK7 may act as an antioxidant and/or a molecular chaperone. It may also detoxify aggressive metabolic aldehyde by-products. It helps in stabilizing mitochondria during cellular stress. Alteration in gene function affects neurodegeneration, which leads to the autosomal recessive early-onset parkinsonism.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Larissa Berloffa Belardin et al.
Journal of assisted reproduction and genetics, 33(12), 1657-1664 (2016-09-16)
To verify if the presence of varicocele (grades II and III) with and without seminal alterations, using the 5 This cross sectional study was performed at the Sao Paulo Federal University Paulo between May 2014 and April 2016. A total
L Belardin et al.
Andrology, 7(1), 53-61 (2018-10-26)
Cysteine-rich secretory protein (CRISP-3), a protein involved in inflammatory response, is highly increased in seminal plasma of adolescents with varicocoele and altered semen analysis, but not in adolescents with varicocoele and normal semen. It is not known, however, whether this
P T Del Giudice et al.
Andrology, 4(3), 447-455 (2016-04-12)
The goal of this study was to determine seminal plasma biomarkers of testicular function in adolescents with varicocoele and to verify enriched gene ontology terms associated to these differential proteomes. An observational study was carried out in an academic research
Yusuke Toyoda et al.
Biology open, 3(8), 777-784 (2014-07-27)
Parkinson's disease is associated with mitochondrial decline in dopaminergic neurons of the substantia nigra. One of the genes linked with the onset of Parkinson's disease, DJ-1/PARK7, belongs to a novel glyoxalase family and influences mitochondrial activity. It has been assumed
Vincenzo Bonifati et al.
Science (New York, N.Y.), 299(5604), 256-259 (2002-11-26)
The DJ-1 gene encodes a ubiquitous, highly conserved protein. Here, we show that DJ-1 mutations are associated with PARK7, a monogenic form of human parkinsonism. The function of the DJ-1 protein remains unknown, but evidence suggests its involvement in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service