Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB2500375

Sigma-Aldrich

Anti-FABP2 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FABPI, Anti-Fatty acid binding protein 2, Anti-I-FABP, Anti-MGC133132

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, canine

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

Gene Information

human ... FABP2(2169)

Immunogen

Peptide with sequence C-EGVEAKRIFKKD from the C Terminus of the protein sequence according to NP_000125.1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luís Felipe Tsuyoshi Mitidiero et al.
Acta cirurgica brasileira, 29 Suppl 2, 43-49 (2014-09-18)
To determine the expression of hepatic L-FABP and intestinal I-FABP in an experimental model of necrotizing enterocolitis (NEC) in neonatal rats. Newborn Sprague-Dawley rats were divided into four groups: Control (C1) - exclusive breastfeeding at the first and sixth procedures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service