Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2500154

Sigma-Aldrich

Anti-BCL2 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-B-cell CLL/lymphoma 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

species reactivity (predicted by homology)

mouse, rat, canine

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCL2(596)

Immunogen

Peptide with sequence AGRTGYDNREIVMKYC from the N Terminus of the protein sequence according to NP_000624; NP_000648.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Russo et al.
Chemico-biological interactions, 305, 79-85 (2019-04-03)
Melanoma is a highly invasive cancer that resists most conventional treatments. Therefore, there is an urgent need to identify alternative anticancer agents able to affect new molecular targets. Drimys winteri (Winteraceae) is a medicinal plant, employed in Brazil and many
Alejandro Madrid et al.
International journal of molecular sciences, 16(4), 7944-7959 (2015-04-11)
With the aim of identifying novel agents with antigrowth and pro-apoptotic activity on melanoma cancer, the present study was undertaken to investigate the biological activity of the resinous exudate of aerial parts from Psoralea glandulosa, and its active components (bakuchiol
Alessandra Russo et al.
Phytotherapy research : PTR, 33(12), 3242-3250 (2019-09-07)
Isocordin 1 and a series of 4-oxyalkyl-isocordoin analogues 2-8 were evaluated for their cytotoxicity effect against human melanoma cells (A2058). Analogues 4, 5, and 6 showed a higher inhibitory activity with IC50 values of 12.91 ± 0.031, 24.88 ± 0.013, and 11.62 ± 0.017, respectively. These
Alessandra Russo et al.
International journal of molecular sciences, 19(1) (2018-01-20)
Prostate cancer is one of the most common forms of cancer in men, and research to find more effective and less toxic drugs has become necessary. In the frame of our ongoing program on traditionally used Salvia species from the
Adriana Carol Eleonora Graziano et al.
Oncotarget, 8(8), 13917-13931 (2017-01-21)
Wilms' tumor gene 1 (WT1) plays complex roles in tumorigenesis, acting as tumor suppressor gene or an oncogene depending on the cellular context. A high WT1 expression level was described in various types of human bone and soft-tissue sarcomas, including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service