Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2500052

Sigma-Aldrich

Anti-AKAP9/AKAP450/CG-NAP antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti- PRKA9, Anti-AKAP350, Anti-CG-NAP, Anti-Hyperion, Anti-YOTIAO

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKAP9(10142)

General description

The gene encoding A-kinase anchoring protein 9 (AKAP9) is localized to human chromosome 7q21–q22. It is a 350-450kDa protein present in the centrosomes and the Golgi apparatus.

Immunogen

Peptide with sequence C-SGSTTQFHAGMR from the C Terminus of the protein sequence according to NP_671700.1; NP_005742.4; NP_671714.1.

Biochem/physiol Actions

A-kinase anchoring protein 9 (AKAP9) is used by cells to anchor the regulatory subunit of type II cAMP-dependent protein kinase A. It coordinates multiple intracellular signaling events and scaffolds protein kinases and phosphatases at the centrosome and the Golgi apparatus. The protein interacts with cdc42 interacting protein 4 (CIP4) at the Golgi apparatus and functions in the maintenance of normal structure of Golgi apparatus.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raffaele Ciampi et al.
The Journal of clinical investigation, 115(1), 94-101 (2005-01-05)
Genes crucial for cancer development can be mutated via various mechanisms, which may reflect the nature of the mutagen. In thyroid papillary carcinomas, mutations of genes coding for effectors along the MAPK pathway are central for transformation. BRAF point mutation
Brent T Steadman et al.
The Journal of biological chemistry, 277(33), 30165-30176 (2002-05-17)
AKAP350 is a multiply spliced family of 350-450-kDa protein kinase A-anchoring proteins localized to the centrosomes and the Golgi apparatus. Using AKAP350A as bait in a yeast two-hybrid screen of a rabbit parietal cell library, we have identified a novel
Ryan A Shanks et al.
The Journal of biological chemistry, 277(43), 40973-40980 (2002-08-07)
AKAP350 can scaffold a number of protein kinases and phosphatases at the centrosome and the Golgi apparatus. We performed a yeast two-hybrid screen of a rabbit parietal cell library with a 3.2-kb segment of AKAP350 (nucleotides 3611-6813). This screen yielded
M Cecilia Larocca et al.
Molecular biology of the cell, 15(6), 2771-2781 (2004-03-30)
The A kinase anchoring protein 350 (AKAP350) is a multiply spliced type II protein kinase A anchoring protein that localizes to the centrosomes in most cells and to the Golgi apparatus in epithelial cells. In the present study, we sought

Articles

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service