SAB2108945
Anti-EIF3I (C-terminal) antibody produced in rabbit HRP conjugated
affinity isolated antibody
Synonym(s):
EIF3S2, PRO2242, TRIP-1, TRIP1, eIF3-beta
Select a Size
$460.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$460.00
Notify Me
Get notified when this item is ready to ship via email.
biological source
rabbit
conjugate
peroxidase conjugate
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
36 kDa
species reactivity (predicted by homology)
rabbit, human, horse, rat, guinea pig, canine, bovine, mouse
concentration
0.5 mg/mL
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 4-8 μg/mL
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... EIF3I(8668)
General description
Sequence
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service