Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2108819

Sigma-Aldrich

Anti-TM6SF2 (C-terminal) antibody produced in rabbit biotin conjugated

affinity isolated antibody

Synonym(s):

CLU1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$460.00

$460.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$460.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.45

$460.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity (predicted by homology)

mouse, human, rabbit, canine, guinea pig, bovine, rat, horse

concentration

0.5 mg/mL

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TM6SF2(53345)

Immunogen

Synthetic peptide directed towards the C-terminal region of Human TM6SF2

Sequence

Synthetic peptide located within the following region: FFTLFRGLVVLDCPTDACFVYIYQYEPYLRDPVAYPKVQMLMYMFYVLPF

Physical form

Supplied at 0.5 mg/ml in phosphate-buffered saline, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service