Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2108715

Sigma-Aldrich

Anti-SLC39A4

affinity isolated antibody

Synonym(s):

Anti- AWMS2, Anti- FLJ20327, Anti- MGC74741, Anti- ZIP4, Anti-AEZ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

66 kDa

species reactivity

rat, human

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable

accession no.

NM_017767

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC39A4(55630)

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC39A4

Biochem/physiol Actions

SLC39A4 is a member of the zinc/iron-regulated transporter-like protein (ZIP) family. The transmembrane protein is required for zinc uptake in the intestine. Mutations in the gene encoding SLC39A4 result in acrodermatitis enteropathica, a rare inherited defect in the absorption of dietary zinc.This gene encodes a member of the zinc/iron-regulated transporter-like protein (ZIP) family. The transmembrane protein is required for zinc uptake in the intestine. Mutations in this gene result in acrodermatitis enteropathica, a rare inherited defect in the absorption of dietary zinc. Multiple transcript variants encoding different isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: PQCLSVEDALGLGEPEGSGLPPGPVLEARYVARLSAAAVLYLSNPEGTCE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service