Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB2108535

Sigma-Aldrich

Anti-NONO (AB2)

IgG fraction of antiserum

Synonym(s):

Anti- NMT55, Anti- NRB54, Anti- P54NRB, Anti-P54

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

52 kDa

species reactivity

human, bovine, guinea pig, dog, rabbit, horse, rat

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

accession no.

NM_007363

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NONO(4841)

Immunogen

Synthetic peptide directed towards the C terminal region of human NONO

Biochem/physiol Actions

NONO is DNA- and RNA binding protein, involved in several nuclear processes. It binds the conventional octamer sequence in double stranded DNA. It also binds single-stranded DNA and RNA at a site independent of the duplex site. It is involved in pre-mRNA splicing and interacts with U5 snRNA. The SFPQ-NONO heteromer associated with MATR3 may play a role in nuclear retention of defective RNAs, be involved in DNA unwinding by modulating the function of topoisomerase I/TOP1 and be involved in DNA nonhomologous end joining (NHEJ) required for double-strand break repair and V(D)J recombination and may stabilize paired DNA ends. NONO binds to an enhancer element in long terminal repeats of endogenous intracisternal A particles (IAPs) and activates transcription.

Sequence

Synthetic peptide located within the following region: DGTLGLTPPTTERFGQAATMEGIGAIGGTPPAFNRAAPGAEFAPNKRRRY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service