Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2108364

Sigma-Aldrich

Anti-Rgs2 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

24kDa

species reactivity

rat, mouse, human, bovine, sheep

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... Rgs2(19735)

Immunogen

Synthetic peptide directed towards the N terminal of human Rgs2

Biochem/physiol Actions

Rgs2 inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. It may play a role in leukemogenesis.

Sequence

Synthetic peptide located within the following region: NSSAPGKPKTGKKSKQQTFIKPSPEEAQLWAEAFDELLASKYGLAAFRAF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manxi Jiang et al.
Molecular reproduction and development, 86(1), 88-99 (2018-11-10)
Embryo implantation is a complicated event that relies on two critical factors: the competent blastocyst and the receptive uterus. Successful implantation results from tight coordination of these two factors. The maternal hormone environment of the uterus and molecular cross-talk between

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service