Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB2108253

Sigma-Aldrich

Anti-VISA antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VISA(57506)

General description

The previously assigned protein identifier Q2HWT5 has been merged into Q7Z434. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the N terminal region of human VISA

Biochem/physiol Actions

Double-stranded RNA viruses are recognized in a cell type-dependent manner by the transmembrane receptor TLR3 or by the cytoplasmic RNA helicases MDA5 and RIGI (ROBO3). These interactions initiate signaling pathways that differ in their initial steps but converge in the activation of the protein kinases IKKA (CHUK) and IKKB (IKBKB), which activate NFKB, or TBK1 and IKKE (IKBKE), which activate IRF3. Activated IRF3 and NFKB induce transcription of IFNB (IFNB1). For the TLR3 pathway, the intermediary molecule before the pathways converge is the cytoplasmic protein TRIF (TICAM1). For RIGI, the intermediary protein is mitochondria-bound VISA. Double-stranded RNA viruses are recognized in a cell type-dependent manner by the transmembrane receptor TLR3 (MIM 603029) or by the cytoplasmic RNA helicases MDA5 (MIM 606951) and RIGI (ROBO3; MIM 608630). These interactions initiate signaling pathways that differ in their initial steps but converge in the activation of the protein kinases IKKA (CHUK; MIM 600664) and IKKB (IKBKB; MIM 603258), which activate NFKB (see MIM 164011), or TBK1 (MIM 604834) and IKKE (IKBKE; MIM 605048), which activate IRF3 (MIM 603734). Activated IRF3 and NFKB induce transcription of IFNB (IFNB1; MIM 147640). For the TLR3 pathway, the intermediary molecule before the pathways converge is the cytoplasmic protein TRIF (TICAM1; MIM 607601). For RIGI, the intermediary protein is mitochondria-bound IPS1 (Sen and Sarkar, 2005 [PubMed 16239922]).[supplied by OMIM]. Sequence Note: This RefSeq record was created from transcript and genomic sequence data because no single transcript was available for the full length of the gene. The extent of this transcript is supported by transcript alignments. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: ETQAPESPGENSEQALQTLSPRAIPRNPDGGPLESSSDLAALSPLTSSGH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service