Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB2108139

Sigma-Aldrich

Anti-FYN antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$493.00

$493.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$493.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$493.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54kDa

species reactivity

guinea pig, rabbit, dog, mouse, bovine, horse, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FYN(2534)

General description

Tyrosine-protein kinase (FYN) is located on human chromosome 6q21. It is a 59 kDa protein with the attachment sites for saturated fatty acid addition in the N terminal, a unique region, a Src-homology 3 (SH3) domain, a Src-homology 2 (SH2) domain, a tyrosine kinase domain (SH1) and a C-terminal negative regulatory domain.

Immunogen

Synthetic peptide directed towards the N terminal region of human FYN

Application

Anti-FYN antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Tyrosine-protein kinase (FYN) is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.

FYN controls mitogenic signals and regulates cell cycle entry and proliferation. Upregulation of FYN in thyroid carcinoma plays an important role in tumorigenesis. Overexpression of FYN in brain is linked to Alzheimer′s disease (AD). It is also involved in T-cell signalling, myelination, learning and memory, neuroinflammation, apoptosis and cell adhesion.

Sequence

Synthetic peptide located within the following region: GCVQCKDKEATKLTEERDGSLNQSSGYRYGTDPTPQHYPSFGVTSIPNYN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pseudomonas aeruginosa infection liberates transmissible, cytotoxic prion amyloids
Balczon R et al.
Faseb Journal, 31(7), 2785-2796 (2017)
Function of the Src-family kinases, Lck and Fyn, in T-cell development and activation
Palacios, Emil H and Weiss, Arthur
Oncogene, 23(48), 7990-7990 (2004)
Regulatory region genetic variation is associated with FYN expression in Alzheimer's disease
Zahratka JA et al.
Neurobiology of Aging, 51, 43-53 (2017)
Upregulation of Tyrosine Kinase FYN in Human Thyroid Carcinoma: Role in Modulating Tumor Cell Proliferation, Invasion, and Migration
Zheng J et al.
Cancer biotherapy & radiopharmaceuticals, 32(9), 320-326 (2017)
Analysis of chromosome 6 deletions in lymphoid malignancies provides evidence for a region of minimal deletion within a 2-megabase segment of 6q21.
Sherratt T, et al.
Chromosome Research, 5(2), 118-124 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service