The immunogen for anti-TM2D3 antibody: synthetic peptide derected towards the C terminal of human TM2D3
Sequence
Synthetic peptide located within the following region: TALALSITLGGFGADRFYLGQWREGLGKLFSFGGLGIWTLIDVLLIGVGY
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.