Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2106136

Sigma-Aldrich

Anti-UBE2O antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-E2-230K, Anti-FLJ12878, Anti-KIAA1734

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

141 kDa

species reactivity

bovine, guinea pig, mouse, rat, horse, human, dog, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBE2O(63893)

General description

The gene UBE2O (ubiquitin conjugating enzyme E2 O) encodes a member of the E2 family of ubiquitin-conjugating enzymes. The encoded protein is large with a molar mass of 140kDa. It contains a ubiquitin-conjugating (UBC) domain and is expressed ubiquitously, with increased expression in brain, skeletal muscle, and heart tissues. It is found to be up-regulated during the reticulocyte stage of erythroid differentiation.

Immunogen

Synthetic peptide directed towards the middle region of human UBE2O

Biochem/physiol Actions

The E2 enzyme encoded by the gene UBE2O (ubiquitin conjugating enzyme E2 O) negatively regulates TRAF6-mediated IL-1R (interleukin-1R)/TLR4 (toll-like receptor4) signaling via the inhibition of polyubiquitination of TRAF6 (tumor necrosis factors receptor associated factor 6). It hinders the formation of MyD88-TRAF6 protein complex. It prevents TRAF6-dependent NF-κB (nuclear factor-κB) signaling. It is involved in the ubiquitination of the nuclear localization signal of BAP1 (BRCA1 associated protein 1), a tumor suppressor that regulates cell proliferation. UBE2O interacts and monoubiquitinates SMAD6, an inhibitory regulator of bone morphogenetic protein (BMP)/SMAD signaling, thereby regulating BMP7 (bone morphogenetic protein)-induced adipogenesis.

Sequence

Synthetic peptide located within the following region: YNEAGFDSDRGLQEGYENSRCYNEMALIRVVQSMTQLVRRPPEVFEQEIR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Autodeubiquitination protects the tumor suppressor BAP1 from cytoplasmic sequestration mediated by the atypical ubiquitin ligase UBE2O
Mashtalir N
Molecular Cell, 54, 392-406 (2014)
Xiaofei Zhang et al.
The EMBO journal, 32(7), 996-1007 (2013-03-05)
SMAD6 is a crucial feedback inhibitory regulator of bone morphogenetic protein (BMP)/SMAD signalling. Although little is known regarding the post-transcriptional modification of inhibitory SMADs and the mechanism by which their function is regulated. In this study, using a whole proteomic
Xiaofei Zhang et al.
Cell research, 23(3), 366-377 (2013-02-06)
Tumor necrosis factor (TNF) receptor-associated factor 6 (TRAF6) is a key regulator of the activation of transcription factor NF-κB by the interleukin-1 receptor (IL-1R)/Toll-like receptor (TLR) superfamily. Recruitment of TRAF6 to the receptor-associated IRAK1-IRAK4-MyD88 adaptor protein complex induces lysine 63

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service