The previously assigned protein identifier Q2HIX7 has been merged into Q96T88. Full details can be found on the UniProt database.
Immunogen
Synthetic peptide directed towards the N terminal region of human UHRF1
Biochem/physiol Actions
This gene encodes a member of a subfamily of RING-finger type E3 ubiquitin ligases. The protein binds to specific DNA sequences, and recruits a histone deacetylase to regulate gene expression. Its expression peaks at late G1 phase and continues during G2
Sequence
Synthetic peptide located within the following region: MGVFAVPPLSADTMWIQVRTMDGRQTHTVDSLSRLTKVEELRRKIQELFH
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.