Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2104161

Sigma-Aldrich

Anti-AKTIP antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FT1, Anti-FTS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

human, rat, bovine, guinea pig, horse, dog, mouse, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKTIP(64400)

General description

AKT interacting protein (AKTIP) is the member of the ubiquitin E2 variant (UEV) enzyme subfamily. It is a sheltering interacting protein. AKTIP gene is located on human chromosome 16q12.2. The protein is homologous E2 variant ubiquitin-conjugating (UEV) enzymes.

Immunogen

Synthetic peptide directed towards the middle region of human AKTIP

Biochem/physiol Actions

AKT interacting protein (AKTIP) is essential for the replication of telomeric DNA. AKTIP plays an important role in lamin related processes, such as maintaining nuclear architecture, telomere homeostasis and cellular senescence It interacts with Akt (non-specific serine/threonine protein kinase) activity and regulates differentiation, proliferation and apoptosis of many cell types. AKTIP helps in regulating radiation cytotoxicity in cervical cancer. It is involved in vesicle trafficking.

Sequence

Synthetic peptide located within the following region: NPSVHDEAREKMLTQKKPEEQHNKSVHVAGLSWVKPGSVQPFSKEEKTVA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CD5- diffuse large B-cell lymphoma with peculiar cyclin D1+ phenotype. Pathologic and molecular characterization of a single case
Lucioni M, et al.
Human Pathology, 42(8), 1204-1208 (2011)
The telomeric protein AKTIP interacts with A-and B-type lamins and is involved in regulation of cellular senescence
Burla R, et al.
Open Biology, 6(8), 160103-160103 (2016)
Fused Toes Homolog modulates radiation cytotoxicity in uterine cervical cancer cells
Anandharaj A, et al.
Molecular Biology Reports, 38(8), 5361-5370 (2011)
AKTIP/Ft1, a new shelterin-interacting factor required for telomere maintenance
Burla R, et al.
PLoS Genetics, 11(6), e1005167-e1005167 (2015)
Sridhar Muthusami et al.
Cancer letters, 351(2), 252-259 (2014-06-28)
The role of Fused Toes Homolog (FTS) in epidermal growth factor (EGF) induced epithelial-mesenchymal transition (EMT) in cervical cancer cells was studied. EGF treatment induced the change of EMT markers and increased cell migration. EGF treatment also increased phosphorylated EGFR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service