SAB2103443
Anti-CSRP1 antibody produced in rabbit
affinity isolated antibody
Synonym(s):
Anti-CRP, Anti-CRP1, Anti-CSRP, Anti-CYRP, Anti-D1S181E
About This Item
Recommended Products
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
20 kDa
species reactivity
rat, human, dog, guinea pig, rabbit, mouse
concentration
0.5 mg - 1 mg/mL
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CSRP1(1465)
General description
Immunogen
Biochem/physiol Actions
LIM domain of CSRP1 is involved in smooth muscle differentiation and protein dimerization. CSRP1 modulates actin filament bundling. It acts as a stress response factor and is involved in growth inhibitory and cytoprotective functions.
Sequence
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service