Synthetic peptide directed towards the middle region of human WNT9B
Biochem/physiol Actions
WNT9B is a ligand for members of the frizzled family of seven transmembrane receptors. WNT9B is a probable developmental protein. WNT9B may be a signaling molecule which affects the development of discrete regions of tissues. WNT9B is likely to signal over only few cell diameters.The WNT gene family consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
Sequence
Synthetic peptide located within the following region: CTCDDSPGLESRQAWQWGVCGDNLKYSTKFLSNFLGSKRGNKDLRARADA
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.