Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2103206

Sigma-Aldrich

Anti-CHCHD4, (N-terminal) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-FLJ31709, Anti-MIA40

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

16 kDa

species reactivity

, bovine, rabbit, human, rat, horse, zebrafish, mouse, chicken, canine

technique(s)

western blot: suitable

immunogen sequence

MSYCRQEGKDRIIFVTKEDHETPSSAELVADDPNDPYEEHGLILPNGNIN

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CHCHD4(131474)

Immunogen

Peptide region of the protein sequence according to NP_001091972.

Physical form

Antibody is lyophilized from PBS buffer with 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service