Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB2103104

Sigma-Aldrich

Anti-GAPDH antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-G3PD, Anti-GAPD, Anti-MGC88685

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

36 kDa

species reactivity

mouse, rat, rabbit, human, horse, pig

technique(s)

western blot: suitable

immunogen sequence

KAGAHLQGGAKRVIISAPSADAPMFVMGVNHEKYDNSLKIISNASCTTNC

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... GAPDH(2597)

Immunogen

Peptide region of the protein sequence according to NP_002037.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Antibody is lyophilized from PBS buffer with 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Fu et al.
Molecular carcinogenesis, 58(11), 2104-2117 (2019-08-31)
Small nucleolar RNA host gene 1 (SNHG1) is critical in the progression of cancers. However, the mechanism by which SNHG1 regulates the progression of colorectal cancer (CRC) remains unclear. Expressions of SNHG1 and miR-137 in CRC tissues and cell lines
Theodore G Drivas et al.
The Journal of clinical investigation, 123(10), 4525-4539 (2013-09-21)
Mutations in the gene centrosomal protein 290 kDa (CEP290) cause an array of debilitating and phenotypically distinct human diseases, ranging from the devastating blinding disease Leber congenital amaurosis (LCA) to Senior-Løken syndrome, Joubert syndrome, and the lethal Meckel-Gruber syndrome. Despite
Gong-Sheng Li et al.
Oncology letters, 14(4), 5013-5017 (2017-11-01)
The present study aimed to investigate the potential effect of leucine-rich repeat containing 3B (LRRC3B) with respect to the inhibition of breast cancer recurrence and metastasis post-anesthesia. The mRNA expression of LRRC3B in breast MDA-MB-231 and MCF-7 cell lines was
Xiaofeng Lei et al.
Molecular medicine reports, 17(6), 8039-8046 (2018-04-26)
Neuroprotective strategies using diazoxide (DZX) have been demonstrated to limit ischemia/reperfusion (I/R)‑induced injury and cell apoptosis. In type 2 diabetes mellitus, DZX has been reported to improve β‑cell function and reduce their apoptosis, through suppressing endoplasmic reticulum (ER) stress. However
Ke Xu et al.
Journal of neurochemistry, 149(6), 799-810 (2018-12-07)
The proportion of major depressive disorder (MDD) patients around the world has increased remarkably. Although many studies of MDD have been conducted based on classic hypotheses, like alteration of the hypothalamic-pituitary-adrenal axis or monoamine neurotransmitters, the mechanisms underlying MDD remain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service