Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2102505

Sigma-Aldrich

Anti-TNPO2 (ab2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ12155, Anti-IPO3, Anti-KPNB2B, Anti-TRN2, Anti-Transportin 2 (importin 3, karyopherin β 2b)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$541.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

100 kDa

species reactivity

human, guinea pig, mouse, rabbit, horse, rat, dog, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNPO2(30000)

General description

The previously assigned protein identifier Q6IN77 has been merged into O14787. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the C terminal region of human TNPO2

Biochem/physiol Actions

Transportin-2 (TNPO2) mediates nuclear import of HuR protein in vitro. It also participates in mRNA export from the nucleus.

Sequence

Synthetic peptide located within the following region: LVQKTLAQAMMYTQHPEQYEAPDKDFMIVALDLLSGLAEGLGGHVEQLVA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service