Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB2100576

Sigma-Aldrich

Anti-DHRS2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Dehydrogenase/reductase (SDR family) member 2, Anti-HEP27

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

30 kDa

species reactivity

human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DHRS2(10202)

General description

The previously assigned protein identifier Q53GS4 has been merged into Q13268. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the middle region of human DHRS2

Biochem/physiol Actions

Hep27, a cell-cycle regulated protein, belongs to the SDR family (short-chain dehydrogenase/reductase family). Hep27 is a NADPH-dependent dicarbonyl reductase enzyme active on xenobiotics.

Sequence

Synthetic peptide located within the following region: LEHCGGVDFLVCSAGVNPLVGSTLGTSEQIWDKILSVNVKSPALLLSQLL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service